missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55165-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to ARHGAP5.
The 34-amino acid tetratricopeptide repeat (TPR) motif is found in a variety of proteins and may mediate protein-protein or protein-membrane interactions.Specifications
| ARHGAP5 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP5 | |
| ARHGAP5, GFI2, p190 B, RhoGAP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11855, 299012, 394 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97393, Q13017 | |
| ARHGAP5 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction