missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£342.00 - £470.00
Specifications
| Antigen | ARHGAP24 |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18447290
|
Novus Biologicals
NBP1-84161-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205426
|
Novus Biologicals
NBP1-84161 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP24 Polyclonal specifically detects ARHGAP24 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ARHGAP24 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp564B1162, Filamin-A-associated RhoGAP, FilGAP, FILGAPRCGAP72, FLJ33877, p73, p73RhoGAP, RAC1- and CDC42-specific GTPase-activating protein of 72 kDa, RC-GAP72, Rho GTPase activating protein 24, rho GTPase-activating protein 24, RhoGAP of 73 kDa, Rho-type GTPase-activating protein 24, Sarcoma antigen NY-SAR-88 | |
| ARHGAP24 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8N264 | |
| 83478 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ILNSDTLGNPTNVRNMSWLPNGYVTLRDNKQKEQAGELGQHNRLSTYDNVHQQFSMMNLDDKQSIDSATWSTSSCEISLPENSNSCRSSTTTCPEQDFFGGNFEDPVLDGPPQDDLSHPRDYESKSD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title