missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-84161-25ul
This item is not returnable.
View return policy
Description
ARHGAP24 Polyclonal specifically detects ARHGAP24 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ARHGAP24 | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Q8N264 | |
| ARHGAP24 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ILNSDTLGNPTNVRNMSWLPNGYVTLRDNKQKEQAGELGQHNRLSTYDNVHQQFSMMNLDDKQSIDSATWSTSSCEISLPENSNSCRSSTTTCPEQDFFGGNFEDPVLDGPPQDDLSHPRDYESKSD | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp564B1162, Filamin-A-associated RhoGAP, FilGAP, FILGAPRCGAP72, FLJ33877, p73, p73RhoGAP, RAC1- and CDC42-specific GTPase-activating protein of 72 kDa, RC-GAP72, Rho GTPase activating protein 24, rho GTPase-activating protein 24, RhoGAP of 73 kDa, Rho-type GTPase-activating protein 24, Sarcoma antigen NY-SAR-88 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 83478 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction