missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argonaute 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54932
This item is not returnable.
View return policy
Description
Argonaute 3 Polyclonal specifically detects Argonaute 3 in Human samples. It is validated for Western Blot.
Specifications
| Argonaute 3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AGO3argonaute 3, argonaute3, eIF2C 3, eIF-2C 3, Eukaryotic translation initiation factor 2C 3, eukaryotic translation initiation factor 2C, 3, FLJ12765, hAgo3, MGC86946, protein argonaute-3 | |
| Rabbit | |
| 69 kDa | |
| 100 μL | |
| Primary | |
| Guinea pig: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| B1ALI0 | |
| AGO3 | |
| Synthetic peptides corresponding to EIF2C3(eukaryotic translation initiation factor 2C, 3) The peptide sequence was selected from the N terminal of EIF2C3. Peptide sequence MCEVLDIHNIDEQPRPLTDSHRVKFTKEIKGLKVEVTHCGTMRRKYRVCN. | |
| Affinity purified | |
| RUO | |
| 192669 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction