missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Synthase Mouse anti-Human, Clone: 1G11, Novus Biologicals™
Mouse Monoclonal Antibody
£241.00 - £378.00
Specifications
| Antigen | Argininosuccinate Synthase |
|---|---|
| Clone | 1G11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244503
|
Novus Biologicals
NBP2-59443-50UL |
50 μL |
£241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236911
|
Novus Biologicals
NBP2-59443-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Argininosuccinate Synthase Monoclonal specifically detects Argininosuccinate Synthase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Argininosuccinate Synthase | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5 | |
| ASS1 | |
| IgG2a κ | |
| Protein A purified |
| 1G11 | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 445 | |
| Recombinant human ASS1 (1-412aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title