missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Synthase Mouse anti-Human, Clone: 1G11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59443-100UL
This item is not returnable.
View return policy
Description
Argininosuccinate Synthase Monoclonal specifically detects Argininosuccinate Synthase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Argininosuccinate Synthase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 445 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1G11 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| ASS1 | |
| Recombinant human ASS1 (1-412aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction