missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 11 publications
£221.00 - £459.00
Specifications
| Gene Accession No. | P07824 |
|---|---|
| Gene Symbols | ARG1 |
| Molecular Weight of Antigen | 37 kDa |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18789243
|
Novus Biologicals
NB100-59740 |
£459.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18592561
|
Novus Biologicals
NB100-59740-25ug |
25 μg |
£221.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.Specifications
| P07824 | |
| ARG1 | |
| 37 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title