missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB100-59740-25ug
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.
Specifications
| Arginase 1/ARG1/liver Arginase | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
| Goat | |
| 37 kDa | |
| 25 μg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 0.5 mg/mL | |
| Western Blot 0.05 - 0.1 μg/mL, Simple Western 5 μg/mL, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA 1:128000 | |
| P07824 | |
| ARG1 | |
| Peptide with sequence C-NHKPETDYLKPPK corresponding to C-Terminus according to NP_058830.2. | |
| Affinity Purified | |
| RUO | |
| 383 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction