missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/NOL3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00 - £397.00
Specifications
| Antigen | ARC/NOL3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18584924
|
Novus Biologicals
NBP1-76462-0.025mg |
0.025 mg |
£133.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18537420
|
Novus Biologicals
NBP1-76462 |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARC/NOL3 Polyclonal antibody specifically detects ARC/NOL3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| ARC/NOL3 | |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NP_003937 | |
| 8996 | |
| IgG | |
| Peptide affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS | |
| Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human ARC. The immunogen is located within the first 50 amino acids of ARC. Amino Acid Squence: GNAQERPSETIDRERKR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title