missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/NOL3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76462-0.025mg
This item is not returnable.
View return policy
Description
ARC/NOL3 Polyclonal antibody specifically detects ARC/NOL3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ARC/NOL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human ARC. The immunogen is located within the first 50 amino acids of ARC. Amino Acid Squence: GNAQERPSETIDRERKR | |
| 0.025 mg | |
| Apoptosis | |
| 8996 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| NP_003937 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction