missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33872
This item is not returnable.
View return policy
Description
Aquaporin-3 Polyclonal specifically detects Aquaporin-3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Aquaporin-3 | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q92482 | |
| AQP3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QLMIGCHLEQPPPSNEEENVKLAHVKHKEQI | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 360 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AQP-3, Aquaglyceroporin-3, aquaporin 3, aquaporin 3 (GIL blood group), aquaporin 3 (Gill blood group), aquaporin-3, GIL, Gill blood group | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction