missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-2 Antibody (5B6B12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £435.00
Specifications
| Antigen | Aquaporin-2 |
|---|---|
| Clone | 5B6B12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250833
|
Novus Biologicals
NBP2-61669 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686167
|
Novus Biologicals
NBP2-61669-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aquaporin-2 Monoclonal specifically detects Aquaporin-2 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Aquaporin-2 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
| AQP2 | |
| IgG1 | |
| Protein G purified |
| 5B6B12 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 359 | |
| Purified recombinant fragment of human AQP2 (AA: 149-271) expressed in E. Coli. | |
| Primary | |
| 28.8 kDa |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel