missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-2 Antibody (5B6B12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61669-0.025ml
This item is not returnable.
View return policy
Description
Aquaporin-2 Monoclonal specifically detects Aquaporin-2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Aquaporin-2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
| Mouse | |
| 28.8 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 5B6B12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AQP2 | |
| Purified recombinant fragment of human AQP2 (AA: 149-271) expressed in E. Coli. | |
| Protein G purified | |
| Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 359 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction