Description
- Provides a complete solution for automated microplate washing workflows
- Supports multiple wash protocols using configurable fluid inlet systems
- Enables precise dispense and aspiration control for consistent assay results
- Supports processing of microplates without cell disruption
- Supports multiple washing modes including sequential, continuous, and bottom washing
- Compatible with interchangeable wash heads for different microplate formats
- Provides flexibility to support current and higher-density microplate applications
- Designed for use in microplate-based assay workflows across life science applications
Specifications
Specifications
| Bottles | Wash Bottle (2 L), Waste Bottle (2 L) |
| Dispense Volume | 25 to 3000 μL/well |
| Plate Type | 96 and 384-well Plates |
| Residual Volume | ≤2 μL/well |
| Soak Time | 1 to 600 sec. |
| Type | Plate Washer |
| Wash Cycles | 1 to 250 |
| Wash Volume | 25 to 3000 μL/well |