missing translation for 'onlineSavingsMsg'
Learn More

Labcon APT Automation Pipette Tips for Beckman Biomek Workstations

Product Code. 17131965
Click to view available options
Volume (Metric):
230 μL
Unit Size:
Pack of 4800
This item is not returnable. View return policy

Product Code. 17131965

Brand: Labcon 80176950109

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Automation certified. Fits Beckman™ Biomek Workstations

  • Equivalent to Beckman™ Biomek P250 pipette tip
  • Sterile - Validated Sterilization (ISO 11137) for use in Research and Medical labs
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Autoclavable
  • Made in USA

Specifications

Description APT™ 230 μL Automation Pipette Tips for Beckman™ Biomek Workstations, in 96 Racks, Sterile
Volume (Metric) 230 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Clear
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
For Use With (Application) Automated Liquid Handling
Tip Style Automation
Packaging 960 Tips/Pk., 5 Pk./Cs.
Conductivity No
Disposable Single-use
For Use With (Equipment) Beckman Coulter Biomek Workstations
Graduated No
Product Line APT
Material Polypropylene Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.