Learn More
Aprotinin From Bovine Lung, Thermo Scientific Chemicals
Useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Thermo Scientific™ Aprotinin From Bovine Lung is not effective against papain.
£58.77 - £160.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15835258
|
Thermo Scientific
J11388.MA |
10 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15845258
|
Thermo Scientific
J11388.MB |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15682109
|
Affymetrix USB
J11388MC |
100 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
- Molecular Formula: C284H432N84O79S7
- MDLNumber: MFCD00130541
- CAS: 9087-70-1
RUO – Research Use Only