missing translation for 'onlineSavingsMsg'
Learn More

Aprotinin From Bovine Lung

Product Code. 15835258
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835258 10 mg 10mg
15845258 25 mg 25mg
15682109 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15835258 Supplier Thermo Scientific Supplier No. J11388.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Thermo Scientific™ Aprotinin From Bovine Lung is not effective against papain. | 6511.44 g/mol

A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
  • Molecular Formula: C284H432N84O79S7
  • MDLNumber: MFCD00130541
  • CAS: 9087-70-1
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 to 2 mg/mL (Soluble in 0.01 M HEPES)
For Use With (Application) Protease Inhibition
Molecular Weight (g/mol) 6511.44
Name Aprotinin
pH Range 8
Purification Method Gel filtration
Quantity 10 mg
Source Bovine lung
Storage Requirements 4°C
Packaging Plastic bottle
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.