missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APRIN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | APRIN |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18434062
|
Novus Biologicals
NBP1-90122-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779653
|
Novus Biologicals
NBP1-90122 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APRIN Polyclonal specifically detects APRIN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| APRIN | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Androgen-induced proliferation inhibitorKIAA0979androgen induced inhibitor of proliferation, Androgen-induced prostate proliferative shutoff-associated protein AS3, androgen-induced shutoff 3, AS3APRIN, CG008, FLJ23236, PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae), sister chromatid cohesion protein PDS5 homolog B | |
| PDS5B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Cycle and Replication, DNA Repair | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23047 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GSQRSRKRGHTASESDEQQWPEEKRLKEDILENEDEQNSPPKKGKRGRPPKPLGGGTPKEEPTMKTSKKGSKKKSGPPAPEEEE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title