Learn More
Apramycin sulfate, 96%, Thermo Scientific™
Potent inhibitor of protein synthesis in bacteria
£78.60 - £270.00
Chemical Identifiers
| CAS | 65710-07-8 |
|---|---|
| Molecular Formula | C21H43N5O15S |
| Molecular Weight (g/mol) | 637.66 |
| MDL Number | MFCD06200257 |
| InChI Key | WGLYHYWDYPSNPF-RQFIXDHTSA-N |
| PubChem CID | 3081544 |
| IUPAC Name | (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid |
| SMILES | OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15476029
|
Thermo Scientific Alfa Aesar
J63874.03 |
1g |
£78.60
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15486029
|
Thermo Scientific Alfa Aesar
J63874.06 |
5g |
£270.00
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsPotent inhibitor of protein synthesis in bacteria.Apramycin sulfate is used as a potent inhibitor of protein synthesis in bacteria. It acts as an antibacterial agent. It is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation. It is also essential for clinic in vitro tests known as antimicrobial susceptibility tests to determine their efficacy against certain bacterial species.
Solubility
Soluble in water.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Incompatible with strong oxidizing agents.
Chemical Identifiers
| 65710-07-8 | |
| 637.66 | |
| WGLYHYWDYPSNPF-RQFIXDHTSA-N | |
| (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid |
| C21H43N5O15S | |
| MFCD06200257 | |
| 3081544 | |
| OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O |
Specifications
| 65710-07-8 | |
| 451.6°C (845°F) | |
| MFCD06200257 | |
| 14,752 | |
| WGLYHYWDYPSNPF-RQFIXDHTSA-N | |
| (2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid | |
| 3081544 | |
| 96% |
| White to Yellow | |
| C21H43N5O15S | |
| 823°C | |
| Soluble in water. | |
| OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O | |
| 637.66 | |
| 637.65 | |
| Apramycin sulfate |
Safety and Handling
GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H360
missing translation for 'einecsNumber' : 265-890-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.