missing translation for 'onlineSavingsMsg'
Learn More

Apramycin sulfate, 96%, Thermo Scientific™

Potent inhibitor of protein synthesis in bacteria

Brand:  Thermo Scientific Alfa Aesar J63874.03

Quantity 1g

 View more versions of this product

Product Code. 15476029

  • £78.60 / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potent inhibitor of protein synthesis in bacteria.Apramycin sulfate is used as a potent inhibitor of protein synthesis in bacteria. It acts as an antibacterial agent. It is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation. It is also essential for clinic in vitro tests known as antimicrobial susceptibility tests to determine their efficacy against certain bacterial species.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

65710-07-8
637.66
WGLYHYWDYPSNPF-RQFIXDHTSA-N
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
C21H43N5O15S
MFCD06200257
3081544
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O

Specifications

65710-07-8
1g
C21H43N5O15S
823°C
Soluble in water.
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
637.66
637.65
Apramycin sulfate
White to Yellow
451.6°C (845°F)
MFCD06200257
14,752
WGLYHYWDYPSNPF-RQFIXDHTSA-N
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
3081544
96%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.