missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79969
This item is not returnable.
View return policy
Description
44289 Polyclonal specifically detects 44289 in Human samples. It is validated for Western Blot.
Specifications
| APR3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| apoptosis related protein 3, apoptosis related protein APR-3, apoptosis-related protein 3, APR3, APR-3, APR--3, chromosome 2 open reading frame 28, HSPC013, p18PRO240 | |
| Rabbit | |
| 18 kDa | |
| 100 μL | |
| Apoptosis | |
| 51374 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_057169 | |
| ATRAID | |
| Synthetic peptide directed towards the C terminal of human C2orf28. Peptide sequence VCADGFHGYKCMRQGSFSLLMFFGILGATTLSVSILLWATQRRKAKTS. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Porcine: 87%; Rat: 87%; Mouse: 87%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction