missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeltDoctor™ MeltDoctor™ HRM Master Mix
Contains all components (excluding template and primers) formulated for superior HRM performance across a wide range of genomic targets.
£566.00 - £4360.00
Specifications
Detection Method | HRM |
---|---|
Form | Liquid |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold 360 DNA Polymerase |
Includes: Provided in a convenient 2X mix, containing: AmpliTaq Gold™ 360 DNA Polymerase, MeltDoctor HRM Dye, a dNTP blend including dUTP, Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10075704
|
Applied Biosystems™
4409535 |
50 mL |
£4285.00
Each |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10351235
|
Applied Biosystems™
4415440 |
5 mL |
£566.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209164
|
Applied Biosystems™
4415450 |
10 x 5 mL |
£4360.00
50mL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10584835
|
Applied Biosystems™
4415452 |
5 x 5 mL |
£2365.00
25mL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
High Resolution Melting (HRM) analysis is a post-PCR analysis method used to identify genetic variation in nucleic acid sequences. Unlike some competitive mixes, the MeltDoctor™ HRM Master Mix does not require additional mixing prior to use, and was developed and optimized solely for HRM applications.
- Low background fluorescence
- High brightness in the presence of double-stranded DNA
- Minimal temperature shift of DNA melting due to dye binding
- Thermal stability to tolerate PCR cycling conditions
- No inhibition of polymerase activity, resulting in high PCR efficiency
- A dNTP blend including dUTP, which minimizes carryover contamination by allowing amplicon degradation by uracil DNA glycosylase (UDG) in subsequent PCR reactions
Perform Mutation Scanning:
- HRM analysis can be used to scan for mutations in target genes for the identification of variant samples prior to sequencing analysis
- As a mutation scanning technique, HRM offers significant advantages over conventional methods such as Denaturing High- Performance Liquid Chromatography (DHPLC) and Denaturing Gradient Gel Electrophoresis (DGGE)
Advantages of HRM for mutation scanning
- Low reagent consumption, with little waste: HRM requires only a 20μL PCR reaction for analysis of each sample, eliminating the need for HPLC solvents or DGGE gels
- Simple, fast workflow: no additional instrumentation is required after PCR amplification;A high-resolution melt step can be simply added to the end of the PCR profile for immediate analysis
- Fast optimization: unlike DHPLC, thermal optimization is not required
- Low sample consumption: following HRM analysis, the PCR product can be used directly in a Sanger sequencing reaction
Quality Components- Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results
- AmpliTaq Gold™ 360 DNA Polymerase, a highly purified DNA polymerase that provides hot-start performance, minimizing non-specific product formation and enabling reactions to be set up at room temperature
- MeltDoctor™ HRM Dye, a stabilized form of the fluorescent SYTO™ 9 double-stranded nucleic acid stain developed by Molecular Probes
Order Info
Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice
Specifications
HRM | |
High | |
AmpliTaq Gold 360 DNA Polymerase | |
HRM (High Resolution Melting) Analysis, Mutation Scanning | |
SNPs (Unknown or Numerous), Epigenetic Targets | |
MeltDoctor™ | |
Genotyping |
Liquid | |
qPCR | |
Fast | |
7500 Fast System, 7900HT Fast System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System | |
High Resolution Melting (HRM), SYTO 9 | |
HRM Master Mix | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.