missing translation for 'onlineSavingsMsg'
Learn More

MeltDoctor™ MeltDoctor™ HRM Master Mix

Contains all components (excluding template and primers) formulated for superior HRM performance across a wide range of genomic targets.

Brand:  Applied Biosystems™ 4415452

Code : 50

Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg

 View more versions of this product

Product Code. 10584835

  • £2365.00 / 25mL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

High Resolution Melting (HRM) analysis is a post-PCR analysis method used to identify genetic variation in nucleic acid sequences. Unlike some competitive mixes, the MeltDoctor™ HRM Master Mix does not require additional mixing prior to use, and was developed and optimized solely for HRM applications.

  • Low background fluorescence
  • High brightness in the presence of double-stranded DNA
  • Minimal temperature shift of DNA melting due to dye binding
  • Thermal stability to tolerate PCR cycling conditions
  • No inhibition of polymerase activity, resulting in high PCR efficiency
  • A dNTP blend including dUTP, which minimizes carryover contamination by allowing amplicon degradation by uracil DNA glycosylase (UDG) in subsequent PCR reactions

Perform Mutation Scanning:
  • HRM analysis can be used to scan for mutations in target genes for the identification of variant samples prior to sequencing analysis
  • As a mutation scanning technique, HRM offers significant advantages over conventional methods such as Denaturing High- Performance Liquid Chromatography (DHPLC) and Denaturing Gradient Gel Electrophoresis (DGGE)

Advantages of HRM for mutation scanning
  • Low reagent consumption, with little waste: HRM requires only a 20μL PCR reaction for analysis of each sample, eliminating the need for HPLC solvents or DGGE gels
  • Simple, fast workflow: no additional instrumentation is required after PCR amplification;A high-resolution melt step can be simply added to the end of the PCR profile for immediate analysis
  • Fast optimization: unlike DHPLC, thermal optimization is not required
  • Low sample consumption: following HRM analysis, the PCR product can be used directly in a Sanger sequencing reaction
    Quality Components
    • Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results
    • AmpliTaq Gold™ 360 DNA Polymerase, a highly purified DNA polymerase that provides hot-start performance, minimizing non-specific product formation and enabling reactions to be set up at room temperature
    • MeltDoctor™ HRM Dye, a stabilized form of the fluorescent SYTO™ 9 double-stranded nucleic acid stain developed by Molecular Probes

    Order Info

    Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice

Specifications

Specifications

Components of the MeltDoctor™ HRM Master Mix, provided in a convenient 2X mix, include: AmpliTaq Gold™ 360 DNA Polymerase, MeltDoctor™ HRM Dye, a dNTP blend including dUTP, Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results.

Store at -15 °C to -25 °C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Liquid
qPCR
Fast
7500 Fast System, 7900HT Fast System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
High Resolution Melting (HRM), SYTO 9
HRM Master Mix
Genotyping
DNA (Genomic), cDNA
HRM
High
AmpliTaq Gold 360 DNA Polymerase
HRM (High Resolution Melting) Analysis, Mutation Scanning
SNPs (Unknown or Numerous), Epigenetic Targets
MeltDoctor™
5 x 5 mL
2500 Reactions
55 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.