missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APPL1 Recombinant Rabbit Monoclonal Antibody (23GB1570), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | APPL1 |
|---|---|
| Clone | 23GB1570 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087708
![]() |
Invitrogen™
MA553668 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087729
|
Invitrogen™
MA553669 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APPL1 Recombinant Monoclonal antibody specifically detects APPL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| APPL1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8K3H0, Q9UKG1 | |
| 26060, 290537, 72993 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB1570 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AI585782; AKT2 interactor; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; DIP13 alpha; DIP13A; DIP13alpha; dip13-alpha; Kiaa1428; MODY14; RGD1309388; signaling adaptor protein DIP13alpha | |
| A synthesized peptide derived from human APPL (400-450AA). | |
| Primary | |
| -20°C | |
| APPL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title