missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APPL1 Recombinant Rabbit Monoclonal Antibody (23GB1570), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553669
This item is not returnable.
View return policy
Description
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
Specifications
| APPL1 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K3H0, Q9UKG1 | |
| APPL1 | |
| A synthesized peptide derived from human APPL (400-450AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1570 | |
| Unconjugated | |
| APPL1 | |
| 2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AI585782; AKT2 interactor; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; DIP13 alpha; DIP13A; DIP13alpha; dip13-alpha; Kiaa1428; MODY14; RGD1309388; signaling adaptor protein DIP13alpha | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26060, 290537, 72993 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction