missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL Rabbit anti-Human, Mouse, Rat, Clone: 4M5X9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | APPL |
|---|---|
| Clone | 4M5X9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18300586
|
Novus Biologicals
NBP3-16721-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18338406
|
Novus Biologicals
NBP3-16721-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APPL Monoclonal antibody specifically detects APPL in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| APPL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Neuroscience, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 26060 | |
| IgG | |
| Affinity purified |
| 4M5X9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human APPL1 (Q9UKG1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title