missing translation for 'onlineSavingsMsg'
Learn More

APPL Rabbit anti-Human, Mouse, Rat, Clone: 4M5X9, Novus Biologicals™

Product Code. 18338406 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338406 100 μg 100µL
18300586 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18338406 Supplier Novus Biologicals Supplier No. NBP316721100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

APPL Monoclonal antibody specifically detects APPL in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen APPL
Applications Western Blot
Classification Monoclonal
Clone 4M5X9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human APPL1 (Q9UKG1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26060
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.