missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | APPBP2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
APPBP2 Polyclonal specifically detects APPBP2 in Human samples. It is validated for Western Blot.Specifications
| APPBP2 | |
| Polyclonal | |
| Rabbit | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| amyloid beta precursor protein (cytoplasmic tail) binding protein 2, Amyloid beta precursor protein-binding protein 2, amyloid protein-binding protein 2, APP-BP2, KIAA0228HS.84084, PAT1amyloid beta precursor protein (cytoplasmic tail)-binding protein 2, Protein interacting with APP tail 1 | |
| APPBP2 | |
| Primary | |
| 67 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q92624 | |
| 10513 | |
| IgG | |
| Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title