missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53085
This item is not returnable.
View return policy
Description
APPBP2 Polyclonal specifically detects APPBP2 in Human samples. It is validated for Western Blot.
Specifications
| APPBP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| amyloid beta precursor protein (cytoplasmic tail) binding protein 2, Amyloid beta precursor protein-binding protein 2, amyloid protein-binding protein 2, APP-BP2, KIAA0228HS.84084, PAT1amyloid beta precursor protein (cytoplasmic tail)-binding protein 2, Protein interacting with APP tail 1 | |
| Rabbit | |
| 67 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled watter to a final antibody concentration of 1mg/mL. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q92624 | |
| APPBP2 | |
| Synthetic peptides corresponding to Amyloid precursor protein (cytoplasmic tail) binding protein 2). The peptide sequence was selected from the middle region of Amyloid precursor protein (NP_006371). Peptide sequence: QASKACVVKREFKKAEQLIKHAVYLARDHFGSKHPKYSDTLLDYGFYLLN The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 10513 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction