missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Apoptotic DNA Ladder Kit

Product Code. 10030040 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 tests
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10030040 50 tests 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030040 Supplier Molecular Probes™ Supplier No. B/2300/PB08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Apoptotic DNA Ladder Kit

The ApoTarget™ Quick Apoptotic DNA Ladder Detection Kit provides a simple and rapid procedure for extraction of chromosomal DNA. The procedure prepares DNA for use in the methods that detect DNA fragmentation in apoptotic cells. Unlike other kits requiring 1 to 2 days to finish, this detection method requires only less than 90 minutes to prepare DNA in a single tube without the need for extraction or column steps. DNA fragmentation can be easily visualized by agarose gel electrophoresis. This procedure increases recovery of small fragmented DNA, thereby improving the sensitivity of the assay.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 tests
Format Tube
Product Type Apoptotic DNA Ladder Kit
Content And Storage Contains TE lysis buffer, enzymes A and B, ammonium acetate solution, and DNA suspension buffer. Store kit at -20°C.
Shipping Condition Approved for shipment on wet or dry ice
Product Line ApoTarget
Conjugate Ethidium Bromide
For Use With (Equipment) UV Imager
compliance-icons compliance-icons
Product Identifier
  • Benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.