missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Apoptotic DNA Ladder Kit

Product Code. 10018932
Change view
Click to view available options
Quantity:
50 tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10018932 50 tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10018932 Supplier Molecular Probes™ Supplier No. KHO1021

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Apoptotic DNA Ladder Kit

The ApoTarget™ Quick Apoptotic DNA Ladder Detection Kit provides a simple and rapid procedure for extraction of chromosomal DNA. The procedure prepares DNA for use in the methods that detect DNA fragmentation in apoptotic cells. Unlike other kits requiring 1 to 2 days to finish, this detection method requires only less than 90 minutes to prepare DNA in a single tube without the need for extraction or column steps. DNA fragmentation can be easily visualized by agarose gel electrophoresis. This procedure increases recovery of small fragmented DNA, thereby improving the sensitivity of the assay.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 tests
Format Tube
Product Type Apoptotic DNA Ladder Kit
Content And Storage Contains TE lysis buffer, enzymes A and B, ammonium acetate solution, and DNA suspension buffer. Store kit at -20°C.
Shipping Condition Approved for shipment on wet or dry ice
Product Line ApoTarget™
Conjugate Ethidium Bromide
For Use With (Equipment) UV Imager

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.