missing translation for 'onlineSavingsMsg'
Learn More

Apoptosis Activator 2, Thermo Scientific Chemicals

Product Code. p-7082452
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 15416839

Brand: Thermo Scientific Alfa Aesar J65320.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines

Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79183-19-0
Molecular Formula C15H9Cl2NO2, C15H9Cl2NO2
Synonym 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione

Specifications

Description Orange
Quantity 5 mg
Formula Weight 306.14g/mol
Sensitivity Light sensitive
Product Type Apoptosis Reagent
Content And Storage 2°C to 8°C
For Use With (Application) Activates apoptosis in cells, shows selectivity for cancer cells against normal cell lines
InChI Key KGRJPLRFGLMQMV-UHFFFAOYSA-N
SMILES ClC1=CC=C(CN2C(=O)C(=O)C3=CC=CC=C23)C=C1Cl
IUPAC Name 1-[(3,4-dichlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione
Molecular Weight (g/mol) 306.14
Form Powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.