missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein O Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £341.00
Specifications
| Antigen | Apolipoprotein O |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17910833
|
Invitrogen™
PA5145414 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17979662
![]() |
Invitrogen™
PA5145413 |
200 μL |
£341.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16.Specifications
| Apolipoprotein O | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Pig | |
| Q9BUR5, Q9DCZ4 | |
| 100525438, 68316, 79135 | |
| Recombinant Human APOO protein, Lys26-Lys198 (Accession #Q9BUR5). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; FAM121B; family with sequence similarity 121B; MGC4825; Mic23; MIC26; MICOS complex subunit Mic23; MICOS complex subunit MIC26; My025; MYO25; Protein FAM121B; RGD1565289; UNQ1866/PRO4302 | |
| APOO | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| APOO |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title