missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein O Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145413
This item is not returnable.
View return policy
Description
This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16.
Specifications
| Apolipoprotein O | |
| Polyclonal | |
| Unconjugated | |
| APOO | |
| 0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; FAM121B; family with sequence similarity 121B; MGC4825; Mic23; MIC26; MICOS complex subunit Mic23; MICOS complex subunit MIC26; My025; MYO25; Protein FAM121B; RGD1565289; UNQ1866/PRO4302 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100525438, 68316, 79135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q9BUR5, Q9DCZ4 | |
| APOO | |
| Recombinant Human APOO protein, Lys26-Lys198 (Accession #Q9BUR5). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction