missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein M/ApoM Antibody (SR1390), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | Apolipoprotein M/ApoM |
|---|---|
| Clone | SR1390 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617994
|
Novus Biologicals
NBP3-22196-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664065
|
Novus Biologicals
NBP3-22196-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein M/ApoM Monoclonal antibody specifically detects Apolipoprotein M/ApoM in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| Apolipoprotein M/ApoM | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 55937 | |
| IgG | |
| Affinity purified |
| SR1390 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| apolipoprotein M, apo-M, ApoM, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a | |
| A synthesized peptide derived from human Apolipoprotein M/ApoM (Uniprot #: O95445) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title