missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein M/ApoM Antibody (SR1390), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22196-50ul
This item is not returnable.
View return policy
Description
Apolipoprotein M/ApoM Monoclonal antibody specifically detects Apolipoprotein M/ApoM in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Apolipoprotein M/ApoM | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55937 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1390 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200 | |
| apolipoprotein M, apo-M, ApoM, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a | |
| A synthesized peptide derived from human Apolipoprotein M/ApoM (Uniprot #: O95445) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction