missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Apolipoprotein L4 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18466821
|
Novus Biologicals
NBP2-14302-25ul |
25ul |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18146195
|
Novus Biologicals
NBP2-14302 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein L4 Polyclonal specifically detects Apolipoprotein L4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Apolipoprotein L4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9BPW4 | |
| 80832 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: TSTDQLEALRDILRDITPNVLSFALDFDEATKMIANDVHTLRRSKATVGRPLIAWRYVPINVVETLRTRGAPTRIVRKVARNL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| apolipoprotein L, 4, apolipoprotein L4, Apolipoprotein L-IV, APOLIV, APOL-IV | |
| APOL4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title