missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14302
This item is not returnable.
View return policy
Description
Apolipoprotein L4 Polyclonal specifically detects Apolipoprotein L4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Apolipoprotein L4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9BPW4 | |
| APOL4 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: TSTDQLEALRDILRDITPNVLSFALDFDEATKMIANDVHTLRRSKATVGRPLIAWRYVPINVVETLRTRGAPTRIVRKVARNL | |
| 0.1 mL | |
| metabolism | |
| 80832 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| apolipoprotein L, 4, apolipoprotein L4, Apolipoprotein L-IV, APOLIV, APOL-IV | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction