missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E R2/ApoE R2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£186.00 - £407.00
Specifications
| Antigen | Apolipoprotein E R2/ApoE R2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, SDS-Page reported in scientific literature (PMID 27810638) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18739063
|
Novus Biologicals
NB100-2216SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781254
|
Novus Biologicals
NB100-2216 |
0.1 mL |
£407.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, SDS-Page.Specifications
| Apolipoprotein E R2/ApoE R2 | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, SDS-Page reported in scientific literature (PMID 27810638) | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1 | |
| LRP8 | |
| IgG | |
| Affinity Purified | |
| Human, chicken and mouse. Immunogen has 95% homology to bovine. |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Chicken | |
| Q14114 | |
| 7804 | |
| A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt# Q14114] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title