missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E R2/ApoE R2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-2216
This item is not returnable.
View return policy
Description
Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, SDS-Page.
Specifications
| Apolipoprotein E R2/ApoE R2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, chicken and mouse. Immunogen has 95% homology to bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, SDS-Page reported in scientific literature (PMID 27810638) | |
| Q14114 | |
| LRP8 | |
| A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963). [UniProt# Q14114] | |
| 0.1 mL | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 7804 | |
| Human, Mouse, Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction