Learn More
Invitrogen™ Apolipoprotein C3 Human ELISA Kit
Sandwich ELISA Kit
£540.00 - £4280.00
Specifications
Accession Number | P02656 |
---|---|
Assay Sensitivity | 2.5 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15444903
|
Invitrogen™
EHAPOC3 |
96 Tests |
£540.00
1 set |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17844180
|
Invitrogen™
EHAPOC3X10 |
10 x 96 Tests |
£4280.00
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17195349
|
Invitrogen™
EHAPOC3X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The Human Apolipoprotein C3 (Hu ApoC3) ELISA quantitates Hu ApoC3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoC3. Principle of the method The Human ApoC3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Apolipoprotein C3 (Apo C3) is produced primarily in the liver, and is known to circulate in plasma in combination with Apo B lipoprotein and high density lipoproteins. Apo C3 acts as an antagonist to Apo C2 and Apo E to prevent lipolysis by the lipoprotein lipase enzyme, and also the clearance of apo B. The gene encoding Apo C3 is found on chromosome 11.Specifications
P02656 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
345 | |
<12% | |
HRP | |
Cardiovascular Research | |
APOCIII, HALP2 | |
1 hr. 20 min. |
2.5 pg/mL | |
Biotin | |
GCPR | |
Human | |
Colorimetric Microplate Reader | |
APOCIII,HALP2 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |