Learn More
Invitrogen™ Apolipoprotein C3 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHAPOC3X5
Additional Details : Weight : 0.33820kg
Description
The Human Apolipoprotein C3 (Hu ApoC3) ELISA quantitates Hu ApoC3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoC3. Principle of the method The Human ApoC3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Apolipoprotein C3 (Apo C3) is produced primarily in the liver, and is known to circulate in plasma in combination with Apo B lipoprotein and high density lipoproteins. Apo C3 acts as an antagonist to Apo C2 and Apo E to prevent lipolysis by the lipoprotein lipase enzyme, and also the clearance of apo B. The gene encoding Apo C3 is found on chromosome 11.Specifications
P02656 | |
2.5 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
345 | |
<12% | |
Chromogen, Pre-coated 96 well plates, Assay Diluent Concentrate, SAV-HRP, Biotinylated Detection Antibody, Standard, Adhesive Plate Covers, Stop Solution, Wash Buffer | |
5 x 96 Tests | |
Cardiovascular Research | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.44 pg/mL to 10,000 pg/mL | |
2.5 pg/mL to 10,000 pg/mL | |
Biotin | |
GCPR | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Alpha-2 Microglobulin, ApoA, ApoC1, ApoC2, ApoE, ApoH, Artemin, BMP-6, Calbindin, CK beta 8-1 (CCL23), CNTF, EMMPRIN (CD147), Endostatin, Fibrinogen, FLRG, TNFSF18 (GITR Ligand), GPBB, Hemoglobin, HTRA2, IL-17 E, IL-1 R3 (IL1 R Acp), Lactoferrin, Mesothelin, PAPP-A, PD-ECGF (Thymidine Phosphorylase), PDGF R alpha, Pepsinogen 2, Periostin, Persephin, PTH, RAP, SAA (Serum Amyloid A), Serpin F1 (PEDF), Tie-1, TPA, Thrombospondin 2, Ubiquitin+1, Vitronectin, VWF | |
Human | |
Colorimetric Microplate Reader | |
APOCIII,HALP2 | |
<10% | |
HRP | |
RUO | |
Plasma, 2 μ;L; Serum, 2 μ;L; Supernatant, 100 μ;L | |
APOCIII, HALP2 | |
1 hr. 20 min. |