missing translation for 'onlineSavingsMsg'
Learn More

APOBEC3F Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen APOBEC3F
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18208943
View Documents
Novus Biologicals
NBP1-57515
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

APOBEC3F Polyclonal specifically detects APOBEC3F in Human samples. It is validated for Western Blot.
Specifications

Specifications

APOBEC3F
Polyclonal
Rabbit
Q8IUX4
200316
IgG
Western Blot
Unconjugated
RUO
apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F, Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F, ARP8, BK150C2.4.MRNA, DNA dC->dU-editing enzyme APOBEC-3F, EC 3.5.4, EC 3.5.4.-, induced upon T-cell activation, KA6, MGC74891
APOBEC3F
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.