missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | APOBEC3F |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
APOBEC3F Polyclonal specifically detects APOBEC3F in Human samples. It is validated for Western Blot.Specifications
| APOBEC3F | |
| Polyclonal | |
| Rabbit | |
| Q8IUX4 | |
| 200316 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F, Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F, ARP8, BK150C2.4.MRNA, DNA dC->dU-editing enzyme APOBEC-3F, EC 3.5.4, EC 3.5.4.-, induced upon T-cell activation, KA6, MGC74891 | |
| APOBEC3F | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title