missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | APOBEC1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
APOBEC1 Polyclonal specifically detects APOBEC1 in Human samples. It is validated for Western Blot.Specifications
| APOBEC1 | |
| Polyclonal | |
| Rabbit | |
| P41238 | |
| 339 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| APOBEC-1, apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1, Apolipoprotein B mRNA-editing enzyme 1, BEDPC->U-editing enzyme APOBEC-1, CDAR1, EC 3.5.4, EC 3.5.4.-, HEPRapolipoprotein B mRNA editing enzyme complex-1 | |
| APOBEC1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title