missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57268
This item is not returnable.
View return policy
Description
APOBEC1 Polyclonal specifically detects APOBEC1 in Human samples. It is validated for Western Blot.
Specifications
| APOBEC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| APOBEC-1, apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1, Apolipoprotein B mRNA-editing enzyme 1, BEDPC->U-editing enzyme APOBEC-1, CDAR1, EC 3.5.4, EC 3.5.4.-, HEPRapolipoprotein B mRNA editing enzyme complex-1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 339 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P41238 | |
| APOBEC1 | |
| Synthetic peptides corresponding to APOBEC1(apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1) The peptide sequence was selected from the N terminal of APOBEC1. Peptide sequence TSEKGPSTGDPTLRRRIEPWEFDVFYDPRELRKEACLLYEIKWGMSRKIW The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 85%; Pig: 85%; Canine: 78%; Equine: 78%. | |
| Human, Mouse, Rat, Pig, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction