missing translation for 'onlineSavingsMsg'
Learn More

Apixaban PG Ester-II, TRC

Product Code. 30389983
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389983 50 mg 50mg
30407457 10 mg -
30397968 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389983 Supplier Toronto Research Chemicals Supplier No. TRCH62813050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1904628-11-0
Molecular Formula C28H30N4O6
Molecular Weight (g/mol) 518.56
Synonym 1-Hydroxypropan-2-yl 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate
IUPAC Name 1-hydroxypropan-2-yl 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate
SMILES COc1ccc(cc1)n2nc(C(=O)OC(C)CO)c3CCN(C(=O)c23)c4ccc(cc4)N5CCCCC5=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H30N4O6/c1-18(17-33)38-28(36)25-23-14-16-31(20-8-6-19(7-9-20)30-15-4-3-5-24(30)34)27(35)26(23)32(29-25)21-10-12-22(37-2)13-11-21/h6-13,18,33H,3-5,14-17H2,1-2H3
Formula Weight 518.2165
Chemical Name or Material Apixaban PG Ester-II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.