CAS RN 1904628-11-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Apixaban PG Ester-II, TRC
CAS: 1904628-11-0 Molecular Formula: C28H30N4O6 Molecular Weight (g/mol): 518.56 Synonym: 1-Hydroxypropan-2-yl 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate IUPAC Name: 1-hydroxypropan-2-yl 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate SMILES: COc1ccc(cc1)n2nc(C(=O)OC(C)CO)c3CCN(C(=O)c23)c4ccc(cc4)N5CCCCC5=O