missing translation for 'onlineSavingsMsg'
Learn More

Apiin, TRC

Product Code. 30138376
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138376 1 mg 1mg
30004055 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138376 Supplier Toronto Research Chemicals Supplier No. TRCA7265501MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26544-34-3
Molecular Formula C26 H28 O14
Molecular Weight (g/mol) 564.49
Synonym Apigenin 7-O-β-D-apiofuranosyl(1→2)-β-D-glucopyranoside, 7-[(2-O-D-apio-β-D-furanosyl-β-D-glucopyranosyl)oxy]-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-Benzopyran-4-one;
IUPAC Name 7-[(2S,3R,4S,5S,6R)-3-[(2S,3S,4R)-3,4-dihydroxy-4-(hydroxymethyl)oxolan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4)[C@H](O[C@@H]5OC[C@](O)(CO)[C@@H]5O)[C@@H](O)[C@@H]1O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H28O14/c27-8-18-20(32)21(33)22(40-25-23(34)26(35,9-28)10-36-25)24(39-18)37-13-5-14(30)19-15(31)7-16(38-17(19)6-13)11-1-3-12(29)4-2-11/h1-7,18,20-25,27-30,32-35H,8-10H2/t18-,20-,21+,22-,23-,24-,25+,26-/m1/s1
Formula Weight 564.1479
Chemical Name or Material Apiin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.