missing translation for 'onlineSavingsMsg'
Learn More

Aphidicolin, Thermo Scientific Chemicals

Product Code. 15443039
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443039 5 mg 5mg
15453039 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443039 Supplier Thermo Scientific Alfa Aesar Supplier No. J60236.#0
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A DNA polymerase inhibitor, blocks the cell cycle at the early S-phase | CAS: 38966-21-1 | C20H34O4, C20H34O4 | 338.49 g/mol

A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Synonym APC

Specifications

Quantity 5 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
Formula Weight 338.48
Melting Point 224°C
Solubility Soluble in DMSO or methanol. Insoluble in water. Store solutions at -20°
Product Type Aphidicolin
Form Cyrstalline powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.